The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080435.2 | NP_001073904.1 | 9 - 10 | 2218 | 66 |
XM_005272421.4 | XP_005272478.1 | 9 - 10 | 1834 | 66 | |
XM_005272422.3 | XP_005272479.1 | 9 - 10 | 1581 | 66 | |
XM_005272423.4 | XP_005272480.1 | 9 - 10 | 1820 | 66 | |
XM_011521232.2 | XP_011519534.1 | 7 - 8 | 2030 | 66 | |
GenBank mRNA | AB075851.1 | - | 9 - 10 | 2204 | 66 |
AK126887.1 | - | 9 - 10 | 2220 | 66 |