Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AAR2
Entrez Gene ID
25980
Gene Name
AAR2 splicing factor homolog
Gene Aliases
C20orf4, CGI-23
Chromosome Location
Chr.20: 36236136 - 36256941 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271874.1 NP_001258803.1 1 - 2 87 109

View Full Product Details