Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAPRE2
Entrez Gene ID
10982
Gene Name
microtubule associated protein RP/EB family member 2
Gene Aliases
CSCSC2, EB1, EB2, RP1
Chromosome Location
Chr.18: 34976928 - 35143470 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143826.2 NP_001137298.1 2 - 3 207 64
NM_001143827.2 NP_001137299.1 2 - 3 353 64
GenBank mRNA AK294833.1 - 2 - 3 297 64
DC335558.1 - 2 - 3 355 64
DC341673.1 - 2 - 3 207 64

View Full Product Details