The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058237.1 | NP_478144.1 | 13 - 14 | 1587 | 67 |
XM_005267930.2 | XP_005267987.1 | 13 - 14 | 1420 | 67 | |
XM_011537038.2 | XP_011535340.1 | 13 - 14 | 1583 | 67 | |
XM_011537039.2 | XP_011535341.1 | 12 - 13 | 1328 | 67 | |
XM_011537040.2 | XP_011535342.1 | 11 - 12 | 1235 | 67 | |
XM_017021528.1 | XP_016877017.1 | 11 - 12 | 1272 | 67 | |
XM_017021529.1 | XP_016877018.1 | 15 - 16 | 1942 | 67 | |
XM_017021530.1 | XP_016877019.1 | 15 - 16 | 1916 | 67 | |
GenBank mRNA | AB046842.1 | - | 13 - 14 | 1506 | 67 |
AK303722.1 | - | 1 - 2 | 69 | 67 | |
BC068491.1 | - | 13 - 14 | 1546 | 67 |