The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_058237.1 | NP_478144.1 | 14 - 15 | 1759 | 98 |
XM_005267930.2 | XP_005267987.1 | 14 - 15 | 1592 | 98 | |
XM_011537038.2 | XP_011535340.1 | 14 - 15 | 1755 | 98 | |
XM_011537039.2 | XP_011535341.1 | 13 - 14 | 1500 | 98 | |
XM_011537040.2 | XP_011535342.1 | 12 - 13 | 1407 | 98 | |
XM_017021528.1 | XP_016877017.1 | 12 - 13 | 1444 | 98 | |
XM_017021529.1 | XP_016877018.1 | 16 - 17 | 2114 | 98 | |
XM_017021530.1 | XP_016877019.1 | 16 - 17 | 2088 | 98 | |
GenBank mRNA | AB046842.1 | - | 14 - 15 | 1678 | 98 |
AK303722.1 | - | 2 - 3 | 241 | 98 | |
BC068491.1 | - | 14 - 15 | 1718 | 98 |