The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020958.2 | NP_066009.2 | 4 - 5 | 539 | 149 |
XM_017021529.1 | XP_016877018.1 | 4 - 5 | 523 | 149 | |
XM_017021530.1 | XP_016877019.1 | 4 - 5 | 497 | 149 | |
GenBank mRNA | BC002650.2 | - | 4 - 5 | 513 | 149 |