The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000615.6 | NP_000606.3 | 16 - 17 | 2467 | 107 |
NM_001242607.1 | NP_001229536.1 | 18 - 19 | 2575 | 107 | |
NM_181351.4 | NP_851996.2 | 17 - 18 | 2497 | 107 | |
GenBank mRNA | AK057509.1 | - | 16 - 17 | 2570 | 107 |
AK292453.1 | - | 18 - 19 | 2132 | 107 | |
AK299865.1 | - | 7 - 8 | 678 | 107 | |
AK314589.1 | - | 18 - 19 | 2547 | 107 | |
BC047244.1 | - | 17 - 18 | 2309 | 107 | |
S71824.1 | - | 16 - 17 | 2302 | 107 | |
U63041.1 | - | 16 - 17 | 2138 | 107 |