Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCAM1
Entrez Gene ID
4684
Gene Name
neural cell adhesion molecule 1
Gene Aliases
CD56, MSK39, NCAM
Chromosome Location
Chr.11: 112961247 - 113278436 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK057509.1 - 18 - 19 2898 58

View Full Product Details