The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000615.6 | NP_000606.3 | 5 - 6 | 999 | 85 |
NM_001076682.3 | NP_001070150.1 | 5 - 6 | 999 | 85 | |
NM_001242607.1 | NP_001229536.1 | 5 - 6 | 999 | 85 | |
NM_001242608.1 | NP_001229537.1 | 5 - 6 | 999 | 85 | |
NM_181351.4 | NP_851996.2 | 5 - 6 | 999 | 85 | |
GenBank mRNA | AB209443.1 | - | 5 - 6 | 878 | 85 |
AK057509.1 | - | 5 - 6 | 1102 | 85 | |
AK292453.1 | - | 5 - 6 | 634 | 85 | |
AK314589.1 | - | 5 - 6 | 971 | 85 | |
BC029119.1 | - | 5 - 6 | 810 | 85 | |
BC047244.1 | - | 5 - 6 | 811 | 85 | |
S71824.1 | - | 5 - 6 | 834 | 85 | |
U63041.1 | - | 5 - 6 | 85 | ||
X16841.1 | - | 5 - 6 | 778 | 85 |