Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NADK
Entrez Gene ID
65220
Gene Name
NAD kinase
Gene Aliases
dJ283E3.1
Chromosome Location
Chr.1: 1751232 - 1780514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198993.1 NP_001185922.1 10 - 11 1250 76
NM_001198994.1 NP_001185923.1 12 - 13 1737 76
NM_001198995.1 NP_001185924.1 8 - 9 1112 76
NM_023018.4 NP_075394.3 10 - 11 1319 76
XM_005244778.2 XP_005244835.1 10 - 11 1380 76
XM_006710837.2 XP_006710900.1 11 - 12 1495 76
XM_006710838.2 XP_006710901.1 10 - 11 1228 76
XM_006710839.1 XP_006710902.1 13 - 14 1807 76
XM_011542006.1 XP_011540308.1 9 - 10 1195 76
XM_011542007.1 XP_011540309.1 9 - 10 1274 76
XM_011542008.1 XP_011540310.1 9 - 10 1132 76
XM_011542009.2 XP_011540311.1 8 - 9 1023 76
XM_017002144.1 XP_016857633.1 8 - 9 1124 76
XM_017002145.1 XP_016857634.1 10 - 11 1563 76
XM_017002146.1 XP_016857635.1 10 - 11 1433 76
GenBank mRNA AF250320.1 - 12 - 13 73
AK023114.1 - 10 - 11 1319 76
AK290161.1 - 10 - 11 1301 76
AK291599.1 - 10 - 11 1243 76
AK291728.1 - 10 - 11 1219 76
AK296722.1 - 8 - 9 1112 76
AY090771.1 - 9 - 10 1097 76
BC001709.1 - 10 - 11 1182 76
BC024219.1 - 3 - 4 407 76

View Full Product Details