Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NADK
Entrez Gene ID
65220
Gene Name
NAD kinase
Gene Aliases
dJ283E3.1
Chromosome Location
Chr.1: 1751232 - 1780514 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198993.1 NP_001185922.1 2 - 3 337 114
NM_001198994.1 NP_001185923.1 2 - 3 389 114
NM_023018.4 NP_075394.3 2 - 3 406 114
XM_005244778.2 XP_005244835.1 2 - 3 467 114
XM_006710837.2 XP_006710900.1 3 - 4 582 114
XM_006710838.2 XP_006710901.1 2 - 3 254 114
XM_006710839.1 XP_006710902.1 3 - 4 582 114
XM_011542006.1 XP_011540308.1 2 - 3 388 114
XM_011542007.1 XP_011540309.1 2 - 3 467 114
XM_011542008.1 XP_011540310.1 2 - 3 325 114
XM_017002143.1 XP_016857632.1 2 - 3 467 114
XM_017002145.1 XP_016857634.1 2 - 3 376 114
XM_017002146.1 XP_016857635.1 2 - 3 246 114
GenBank mRNA AF250320.1 - 2 - 3 389 114
AK023114.1 - 2 - 3 406 114
AK290161.1 - 2 - 3 388 114
AK291599.1 - 2 - 3 330 114
AK291728.1 - 2 - 3 306 114
AK307645.1 - 1 - 2 188 114
AY090771.1 - 1 - 2 184 114
BC001709.1 - 2 - 3 269 114

View Full Product Details