The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198994.1 | NP_001185923.1 | 3 - 4 | 471 | 63 |
XM_006710839.1 | XP_006710902.1 | 4 - 5 | 664 | 63 | |
GenBank mRNA | AF250320.1 | - | 3 - 4 | 471 | 63 |