Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 10 - 11 1343 67
NM_182961.3 NP_892006.3 10 - 11 1487 67
XM_006715407.1 XP_006715470.1 10 - 11 1385 67
XM_006715408.2 XP_006715471.1 10 - 11 1389 67
XM_006715409.1 XP_006715472.1 9 - 10 1364 67
XM_006715410.2 XP_006715473.1 10 - 11 1389 67
XM_006715412.2 XP_006715475.1 10 - 11 1389 67
XM_006715413.2 XP_006715476.1 10 - 11 1389 67
XM_006715415.2 XP_006715478.1 10 - 11 1389 67
XM_006715416.2 XP_006715479.1 10 - 11 1389 67
XM_006715417.2 XP_006715480.1 10 - 11 1389 67
XM_006715420.2 XP_006715483.1 10 - 11 1389 67
XM_006715421.2 XP_006715484.1 10 - 11 1389 67
XM_006715422.1 XP_006715485.1 10 - 11 1385 67
XM_006715423.2 XP_006715486.1 10 - 11 1389 67
XM_006715424.2 XP_006715487.1 10 - 11 1389 67
XM_006715425.2 XP_006715488.1 10 - 11 1389 67
XM_011535641.2 XP_011533943.1 10 - 11 1389 67
XM_011535642.2 XP_011533944.1 10 - 11 1389 67
XM_011535643.1 XP_011533945.1 10 - 11 1385 67
XM_017010608.1 XP_016866097.1 11 - 12 1504 67
XM_017010609.1 XP_016866098.1 10 - 11 1311 67
XM_017010610.1 XP_016866099.1 9 - 10 1290 67
XM_017010611.1 XP_016866100.1 9 - 10 1093 67
XM_017010613.1 XP_016866102.1 10 - 11 1389 67
XM_017010614.1 XP_016866103.1 10 - 11 1389 67
XM_017010615.1 XP_016866104.1 10 - 11 1389 67
XM_017010616.1 XP_016866105.1 10 - 11 1389 67
XM_017010617.1 XP_016866106.1 10 - 11 1389 67
XM_017010618.1 XP_016866107.1 10 - 11 1389 67
GenBank mRNA AF495910.1 - 10 - 11 1487 67
AF535142.1 - 10 - 11 1343 67
AK094094.1 - 7 - 8 804 67
AY183142.1 - 8 - 9 1094 67
JQ740783.1 - 11 - 12 1508 67

View Full Product Details