Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 22 - 23 3024 107
NM_182961.3 NP_892006.3 22 - 23 3168 107
XM_006715407.1 XP_006715470.1 22 - 23 3066 107
XM_006715408.2 XP_006715471.1 22 - 23 3070 107
XM_006715409.1 XP_006715472.1 21 - 22 3045 107
XM_006715410.2 XP_006715473.1 22 - 23 3070 107
XM_006715411.1 XP_006715474.1 21 - 22 3015 107
XM_006715412.2 XP_006715475.1 22 - 23 3070 107
XM_006715413.2 XP_006715476.1 22 - 23 3070 107
XM_006715414.1 XP_006715477.1 20 - 21 2994 107
XM_006715415.2 XP_006715478.1 22 - 23 3070 107
XM_006715416.2 XP_006715479.1 22 - 23 3070 107
XM_006715417.2 XP_006715480.1 22 - 23 3070 107
XM_006715420.2 XP_006715483.1 22 - 23 3070 107
XM_006715421.2 XP_006715484.1 22 - 23 3070 107
XM_006715423.2 XP_006715486.1 22 - 23 3070 107
XM_006715424.2 XP_006715487.1 22 - 23 3070 107
XM_006715425.2 XP_006715488.1 22 - 23 3070 107
XM_011535641.2 XP_011533943.1 22 - 23 3070 107
XM_011535642.2 XP_011533944.1 22 - 23 3070 107
XM_011535643.1 XP_011533945.1 21 - 22 2901 107
XM_011535644.1 XP_011533946.1 8 - 9 1117 107
XM_011535645.2 XP_011533947.1 4 - 5 510 107
XM_017010608.1 XP_016866097.1 23 - 24 3185 107
XM_017010609.1 XP_016866098.1 22 - 23 2992 107
XM_017010610.1 XP_016866099.1 21 - 22 2971 107
XM_017010611.1 XP_016866100.1 21 - 22 2774 107
XM_017010612.1 XP_016866101.1 20 - 21 2698 107
XM_017010613.1 XP_016866102.1 22 - 23 3070 107
XM_017010614.1 XP_016866103.1 22 - 23 3070 107
XM_017010615.1 XP_016866104.1 22 - 23 3070 107
XM_017010616.1 XP_016866105.1 22 - 23 3070 107
XM_017010617.1 XP_016866106.1 22 - 23 3070 107
XM_017010618.1 XP_016866107.1 22 - 23 3070 107
XM_017010619.1 XP_016866108.1 8 - 9 1149 107
GenBank mRNA AB051543.1 - 9 - 10 1241 107
AF495910.1 - 22 - 23 3168 107
AF535142.1 - 22 - 23 3024 107
AK055440.1 - 8 - 9 1191 107
AK308717.1 - 9 - 10 1275 107
AL713682.1 - 3 - 4 340 107
CR933676.1 - 9 - 10 1169 107
FM162565.1 - 20 - 21 2784 107
KU310547.1 - 9 - 10 1272 107
KU310548.1 - 7 - 8 1139 107

View Full Product Details