Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYNE1
Entrez Gene ID
23345
Gene Name
spectrin repeat containing nuclear envelope protein 1
Gene Aliases
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location
Chr.6: 152121684 - 152637399 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033071.3 NP_149062.1 98 - 99 18928 59
NM_182961.3 NP_892006.3 99 - 100 19306 59
XM_006715407.1 XP_006715470.1 99 - 100 19219 59
XM_006715408.2 XP_006715471.1 99 - 100 19211 59
XM_006715409.1 XP_006715472.1 98 - 99 19198 59
XM_006715410.2 XP_006715473.1 99 - 100 19223 59
XM_006715411.1 XP_006715474.1 98 - 99 19168 59
XM_006715412.2 XP_006715475.1 99 - 100 19208 59
XM_006715413.2 XP_006715476.1 99 - 100 19223 59
XM_006715414.1 XP_006715477.1 97 - 98 19147 59
XM_006715415.2 XP_006715478.1 99 - 100 19223 59
XM_006715416.2 XP_006715479.1 99 - 100 19208 59
XM_006715417.2 XP_006715480.1 98 - 99 19082 59
XM_006715420.2 XP_006715483.1 98 - 99 19070 59
XM_006715421.2 XP_006715484.1 98 - 99 19067 59
XM_006715422.1 XP_006715485.1 98 - 99 19060 59
XM_006715423.2 XP_006715486.1 99 - 100 19223 59
XM_006715424.2 XP_006715487.1 99 - 100 19223 59
XM_006715425.2 XP_006715488.1 99 - 100 19223 59
XM_011535641.2 XP_011533943.1 99 - 100 19220 59
XM_011535642.2 XP_011533944.1 99 - 100 19208 59
XM_011535643.1 XP_011533945.1 98 - 99 19054 59
XM_011535644.1 XP_011533946.1 85 - 86 17270 59
XM_011535645.2 XP_011533947.1 81 - 82 16663 59
XM_017010608.1 XP_016866097.1 100 - 101 19338 59
XM_017010609.1 XP_016866098.1 99 - 100 19145 59
XM_017010610.1 XP_016866099.1 98 - 99 19124 59
XM_017010611.1 XP_016866100.1 98 - 99 18927 59
XM_017010612.1 XP_016866101.1 97 - 98 18851 59
XM_017010613.1 XP_016866102.1 99 - 100 19220 59
XM_017010614.1 XP_016866103.1 98 - 99 19067 59
XM_017010615.1 XP_016866104.1 98 - 99 19067 59
XM_017010616.1 XP_016866105.1 99 - 100 19223 59
XM_017010617.1 XP_016866106.1 99 - 100 19220 59
XM_017010618.1 XP_016866107.1 99 - 100 19208 59
XM_017010619.1 XP_016866108.1 85 - 86 17302 59
GenBank mRNA AF495910.1 - 99 - 100 19306 59
AF535142.1 - 98 - 99 18928 59
AY061755.1 - 14 - 15 2396 59
AY184206.1 - 12 - 13 2291 59

View Full Product Details