The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015511.4 | NP_056326.2 | 1 - 2 | 363 | 112 |
GenBank mRNA | BC019311.1 | - | 1 - 2 | 255 | 112 |