The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001322336.1 | NP_001309265.1 | 1 - 2 | 134 | 100 |
NM_001322337.1 | NP_001309266.1 | 1 - 2 | 134 | 100 | |
NM_033411.4 | NP_219479.2 | 1 - 2 | 134 | 100 | |
XM_017010222.1 | XP_016865711.1 | 1 - 2 | 98 | 100 | |
GenBank mRNA | AK309249.1 | - | 1 - 2 | 83 | 100 |
DC377982.1 | - | 1 - 2 | 134 | 100 |