The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002929.2 | NP_002920.1 | 2 - 3 | 935 | 80 |
XM_017020684.1 | XP_016876173.1 | 2 - 3 | 2602 | 80 | |
GenBank mRNA | AK289912.1 | - | 2 - 3 | 1059 | 80 |
L77503.1 | - | 2 - 3 | 842 | 80 | |
U63973.1 | - | 2 - 3 | 935 | 80 |