The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198719.1 | NP_001185648.1 | 4 - 5 | 825 | 135 |
NM_002893.3 | NP_002884.1 | 4 - 5 | 837 | 135 | |
GenBank mRNA | AK091911.1 | - | 4 - 5 | 822 | 135 |
AK291567.1 | - | 4 - 5 | 135 | ||
AK310309.1 | - | 1 - 2 | 126 | 135 | |
BC114500.1 | - | 4 - 5 | 735 | 135 | |
BC114501.1 | - | 4 - 5 | 735 | 135 | |
CR541701.1 | - | 4 - 5 | 479 | 135 | |
U35143.1 | - | 4 - 5 | 740 | 135 | |
X72841.1 | - | 4 - 5 | 764 | 135 |