The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042544.1 | NP_001036009.1 | 11 - 12 | 1512 | 132 |
NM_001042545.1 | NP_001036010.1 | 8 - 9 | 1328 | 132 | |
NM_003573.2 | NP_003564.2 | 11 - 12 | 1401 | 132 | |
XM_011527377.2 | XP_011525679.1 | 11 - 12 | 1560 | 132 | |
XM_011527378.2 | XP_011525680.1 | 11 - 12 | 1560 | 132 | |
XM_011527380.2 | XP_011525682.1 | 11 - 12 | 1560 | 132 | |
XM_011527381.2 | XP_011525683.1 | 11 - 12 | 1560 | 132 | |
XM_011527382.2 | XP_011525684.1 | 11 - 12 | 1560 | 132 | |
XM_011527383.2 | XP_011525685.1 | 11 - 12 | 1560 | 132 | |
XM_011527384.2 | XP_011525686.1 | 11 - 12 | 1560 | 132 | |
XM_011527385.2 | XP_011525687.1 | 11 - 12 | 1560 | 132 | |
XM_011527386.2 | XP_011525688.1 | 11 - 12 | 1560 | 132 | |
XM_011527387.1 | XP_011525689.1 | 11 - 12 | 1660 | 132 | |
XM_017027352.1 | XP_016882841.1 | 11 - 12 | 1560 | 132 | |
XM_017027353.1 | XP_016882842.1 | 11 - 12 | 1560 | 132 | |
XM_017027354.1 | XP_016882843.1 | 11 - 12 | 1560 | 132 | |
GenBank mRNA | Y13622.1 | - | 10 - 11 | 1401 | 132 |