The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042544.1 | NP_001036009.1 | 30 - 31 | 4182 | 161 |
NM_001042545.1 | NP_001036010.1 | 27 - 28 | 3998 | 161 | |
NM_003573.2 | NP_003564.2 | 30 - 31 | 4071 | 161 | |
XM_011527376.2 | XP_011525678.1 | 29 - 30 | 4325 | 161 | |
XM_011527377.2 | XP_011525679.1 | 30 - 31 | 4230 | 161 | |
XM_011527378.2 | XP_011525680.1 | 29 - 30 | 4158 | 161 | |
XM_011527379.1 | XP_011525681.1 | 26 - 27 | 4133 | 161 | |
XM_011527380.2 | XP_011525682.1 | 29 - 30 | 4104 | 161 | |
XM_011527381.2 | XP_011525683.1 | 29 - 30 | 4104 | 161 | |
XM_011527382.2 | XP_011525684.1 | 29 - 30 | 3987 | 161 | |
XM_011527383.2 | XP_011525685.1 | 28 - 29 | 3891 | 161 | |
XM_011527384.2 | XP_011525686.1 | 27 - 28 | 3765 | 161 | |
XM_011527385.2 | XP_011525687.1 | 27 - 28 | 3759 | 161 | |
XM_011527386.2 | XP_011525688.1 | 26 - 27 | 3633 | 161 | |
XM_011527387.1 | XP_011525689.1 | 30 - 31 | 4330 | 161 | |
XM_017027352.1 | XP_016882841.1 | 29 - 30 | 4098 | 161 | |
XM_017027353.1 | XP_016882842.1 | 28 - 29 | 3972 | 161 | |
XM_017027354.1 | XP_016882843.1 | 26 - 27 | 3639 | 161 | |
GenBank mRNA | AF051344.2 | - | 27 - 28 | 3998 | 161 |
AF051345.2 | - | 30 - 31 | 4182 | 161 | |
AF054502.1 | - | 12 - 13 | 1595 | 161 | |
AK074499.1 | - | 8 - 9 | 1486 | 161 | |
AK128191.1 | - | 16 - 17 | 2180 | 161 | |
AK304612.1 | - | 5 - 6 | 908 | 161 | |
Y13622.1 | - | 29 - 30 | 4071 | 161 |