Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DRP2
Entrez Gene ID
1821
Gene Name
dystrophin related protein 2
Gene Aliases
DRP-2
Chromosome Location
Chr.X: 101219786 - 101264497 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171184.1 NP_001164655.1 14 - 15 1951 63
NM_001939.2 NP_001930.2 16 - 17 2234 63
XM_017029333.1 XP_016884822.1 15 - 16 2317 63
XM_017029334.1 XP_016884823.1 15 - 16 2313 63
GenBank mRNA AB209305.1 - 7 - 8 831 63
AK289825.1 - 16 - 17 2122 63
AK295843.1 - 14 - 15 1820 63
BC111695.1 - 16 - 17 2071 63
U43519.1 - 16 - 17 2183 63

View Full Product Details