The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001939.2 | NP_001930.2 | 2 - 3 | 300 | 119 |
XM_017029333.1 | XP_016884822.1 | 2 - 3 | 458 | 119 | |
XM_017029334.1 | XP_016884823.1 | 2 - 3 | 458 | 119 | |
GenBank mRNA | AK289825.1 | - | 2 - 3 | 188 | 119 |
BC111695.1 | - | 2 - 3 | 137 | 119 | |
DA324498.1 | - | 2 - 3 | 301 | 119 |