The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011537.2 | NP_001011537.2 | 4 - 5 | 540 | 130 |
NM_032288.6 | NP_115664.2 | 3 - 4 | 599 | 130 | |
NR_027840.1 | 3 - 4 | 710 | 130 | ||
GenBank mRNA | AJ344094.1 | - | 3 - 4 | 445 | 130 |
AK027672.1 | - | 3 - 4 | 483 | 130 | |
AK302546.1 | - | 3 - 4 | 710 | 130 | |
AK303462.1 | - | 4 - 5 | 540 | 130 | |
AK315019.1 | - | 3 - 4 | 448 | 130 | |
AK316308.1 | - | 3 - 4 | 340 | 130 | |
AL136558.1 | - | 3 - 4 | 452 | 130 | |
BC035006.2 | - | 3 - 4 | 602 | 130 | |
BC039734.1 | - | 3 - 4 | 509 | 130 | |
CR533502.1 | - | 3 - 4 | 380 | 130 |