The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_052997.2 | NP_443723.2 | 1 - 2 | 148 | 89 |
XM_011519757.2 | XP_011518059.1 | 1 - 2 | 251 | 89 | |
XM_011519758.2 | XP_011518060.1 | 1 - 2 | 251 | 89 | |
GenBank mRNA | AF269087.1 | - | 1 - 2 | 148 | 89 |