Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KDM4B
Entrez Gene ID
23030
Gene Name
lysine demethylase 4B
Gene Aliases
JMJD2B, TDRD14B
Chromosome Location
Chr.19: 4969112 - 5153603 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015015.2 NP_055830.1 2 - 3 207 109
XM_005259521.3 XP_005259578.2 2 - 3 208 109
XM_011527818.2 XP_011526120.1 2 - 3 208 109
XM_011527819.1 XP_011526121.1 2 - 3 208 109
XM_011527820.1 XP_011526122.1 2 - 3 208 109
XM_011527821.1 XP_011526123.1 2 - 3 208 109
XM_011527822.1 XP_011526124.1 2 - 3 208 109
XM_017026503.1 XP_016881992.1 2 - 3 208 109
XM_017026504.1 XP_016881993.1 2 - 3 208 109
XM_017026505.1 XP_016881994.1 2 - 3 208 109
GenBank mRNA AB020683.2 - 2 - 3 199 109
BC063889.1 - 2 - 3 188 109
DB636535.1 - 2 - 3 207 109

View Full Product Details