The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300779.1 | NP_001287708.1 | 4 - 5 | 591 | 68 |
NM_015954.3 | NP_057038.2 | 4 - 5 | 591 | 68 | |
GenBank mRNA | AF132960.1 | - | 4 - 5 | 452 | 68 |
AK024614.1 | - | 4 - 5 | 407 | 68 | |
AK222541.1 | - | 4 - 5 | 407 | 68 | |
AK296697.1 | - | 4 - 5 | 348 | 68 | |
AK309182.1 | - | 4 - 5 | 507 | 68 | |
BC041328.1 | - | 4 - 5 | 591 | 68 | |
BC056234.1 | - | 4 - 5 | 433 | 68 | |
BU928832.1 | - | 4 - 5 | 397 | 68 |