The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198505.2 | NP_940907.2 | 16 - 17 | 1915 | 70 |
XM_011512770.1 | XP_011511072.1 | 16 - 17 | 2014 | 70 | |
XM_017006304.1 | XP_016861793.1 | 12 - 13 | 1228 | 70 | |
XM_017006305.1 | XP_016861794.1 | 3 - 4 | 331 | 70 | |
GenBank mRNA | AK122613.1 | - | - | 2035 | 70 |
AY823163.1 | - | 16 - 17 | 1915 | 70 |