Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- CTNNA1
- Entrez Gene ID
-
1495
- Gene Name
- catenin alpha 1
- Gene Aliases
-
CAP102, MDPT2
- Chromosome Location
-
Chr.5: 138753386 - 138935034 on Build GRCh38
View Full Product Details