The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324116.1 | NP_001311045.1 | 8 - 9 | 1756 | 63 |
XM_011509913.2 | XP_011508215.1 | 9 - 10 | 1589 | 63 | |
XM_011509914.2 | XP_011508216.1 | 9 - 10 | 1641 | 63 |