Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HCLS1
Entrez Gene ID
3059
Gene Name
hematopoietic cell-specific Lyn substrate 1
Gene Aliases
CTTNL, HS1, lckBP1, p75
Chromosome Location
Chr.3: 121631398 - 121660948 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001292041.1 NP_001278970.1 10 - 11 1023 84
NM_005335.5 NP_005326.2 11 - 12 1134 84
GenBank mRNA AK124015.1 - 6 - 7 1808 84
AK225633.1 - 10 - 11 1007 84
AK298663.1 - 10 - 11 1058 84
AK298696.1 - 10 - 11 973 84
AK312750.1 - 11 - 12 1097 84
BC016758.2 - 11 - 12 1054 84
BP338037.1 - 4 - 5 435 84
BT006824.1 - 10 - 11 1005 84
CR456794.1 - 10 - 11 1005 84
X16663.1 - 11 - 12 1047 84

View Full Product Details