The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001315501.1 | NP_001302430.1 | 5 - 6 | 968 | 88 |
NM_002555.5 | NP_002546.3 | 5 - 6 | 752 | 88 | |
NM_183233.2 | NP_899056.2 | 5 - 6 | 755 | 88 | |
XM_011520141.2 | XP_011518443.2 | 5 - 6 | 911 | 88 | |
XM_011520142.2 | XP_011518444.2 | 5 - 6 | 910 | 88 | |
GenBank mRNA | AB012083.1 | - | 5 - 6 | 731 | 88 |
AF028738.1 | - | 5 - 6 | 731 | 88 | |
AF030302.1 | - | 5 - 6 | 968 | 88 | |
AF037064.1 | - | 5 - 6 | 739 | 88 | |
AF059663.1 | - | 5 - 6 | 731 | 88 | |
AF070479.1 | - | 5 - 6 | 731 | 88 | |
BC015571.2 | - | 5 - 6 | 755 | 88 | |
BM561526.1 | - | 5 - 6 | 698 | 88 |