The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018023.4 | NP_060493.3 | 18 - 19 | 2778 | 74 |
XM_005247597.2 | XP_005247654.1 | 18 - 19 | 2795 | 74 | |
XM_005247598.4 | XP_005247655.1 | 18 - 19 | 2703 | 74 | |
XM_006713688.3 | XP_006713751.1 | 18 - 19 | 2913 | 74 | |
XM_011512965.1 | XP_011511267.1 | 17 - 18 | 2633 | 74 | |
XM_011512966.1 | XP_011511268.1 | 18 - 19 | 2795 | 74 | |
XM_017006809.1 | XP_016862298.1 | 18 - 19 | 2909 | 74 | |
XM_017006810.1 | XP_016862299.1 | 18 - 19 | 2700 | 74 | |
XM_017006811.1 | XP_016862300.1 | 17 - 18 | 2565 | 74 | |
XM_017006812.1 | XP_016862301.1 | 18 - 19 | 2799 | 74 | |
XM_017006813.1 | XP_016862302.1 | 17 - 18 | 2599 | 74 | |
XM_017006814.1 | XP_016862303.1 | 17 - 18 | 2580 | 74 | |
GenBank mRNA | AB033023.1 | - | 18 - 19 | 2778 | 74 |
BC032368.1 | - | 1 - 2 | 71 | 74 | |
BC043393.1 | - | 4 - 5 | 729 | 74 | |
BC082270.1 | - | 1 - 2 | 83 | 74 | |
BC150273.1 | - | 18 - 19 | 2778 | 74 |