The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018023.4 | NP_060493.3 | 5 - 6 | 730 | 68 |
XM_005247597.2 | XP_005247654.1 | 5 - 6 | 744 | 68 | |
XM_005247598.4 | XP_005247655.1 | 5 - 6 | 652 | 68 | |
XM_006713688.3 | XP_006713751.1 | 5 - 6 | 862 | 68 | |
XM_011512965.1 | XP_011511267.1 | 5 - 6 | 744 | 68 | |
XM_011512966.1 | XP_011511268.1 | 5 - 6 | 744 | 68 | |
XM_017006809.1 | XP_016862298.1 | 5 - 6 | 861 | 68 | |
XM_017006810.1 | XP_016862299.1 | 5 - 6 | 652 | 68 | |
XM_017006811.1 | XP_016862300.1 | 4 - 5 | 514 | 68 | |
XM_017006812.1 | XP_016862301.1 | 5 - 6 | 751 | 68 | |
XM_017006813.1 | XP_016862302.1 | 5 - 6 | 732 | 68 | |
XM_017006814.1 | XP_016862303.1 | 5 - 6 | 641 | 68 | |
GenBank mRNA | AB033023.1 | - | 5 - 6 | 730 | 68 |
AK023370.1 | - | 4 - 5 | 533 | 68 | |
BC150273.1 | - | 5 - 6 | 730 | 68 |