The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018023.4 | NP_060493.3 | 7 - 8 | 1006 | 85 |
XM_005247597.2 | XP_005247654.1 | 7 - 8 | 1020 | 85 | |
XM_005247598.4 | XP_005247655.1 | 7 - 8 | 928 | 85 | |
XM_006713688.3 | XP_006713751.1 | 7 - 8 | 1138 | 85 | |
XM_011512966.1 | XP_011511268.1 | 7 - 8 | 1020 | 85 | |
XM_017006809.1 | XP_016862298.1 | 7 - 8 | 1137 | 85 | |
XM_017006810.1 | XP_016862299.1 | 7 - 8 | 928 | 85 | |
XM_017006811.1 | XP_016862300.1 | 6 - 7 | 790 | 85 | |
XM_017006812.1 | XP_016862301.1 | 7 - 8 | 1027 | 85 | |
XM_017006813.1 | XP_016862302.1 | 7 - 8 | 1008 | 85 | |
GenBank mRNA | AB033023.1 | - | 7 - 8 | 1006 | 85 |
AK023370.1 | - | 6 - 7 | 809 | 85 | |
BC150273.1 | - | 7 - 8 | 1006 | 85 |