Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSL1D1
Entrez Gene ID
26156
Gene Name
ribosomal L1 domain containing 1
Gene Aliases
CSIG, L12, PBK1, UTP30
Chromosome Location
Chr.16: 11834196 - 11851585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015659.2 NP_056474.2 1 - 2 182 71
GenBank mRNA AJ007398.1 - 1 - 2 114 71
AK000822.1 - 1 - 2 126 71
AL049999.1 - 1 - 2 169 71
AY598331.1 - 1 - 2 109 71
BC016362.1 - 1 - 2 145 71
BC017899.1 - 1 - 2 131 71
BC019069.2 - 1 - 2 115 71
BC070325.1 - 1 - 2 133 71
BC107782.1 - 1 - 2 135 71
BC112228.1 - 1 - 2 147 71
BC113699.1 - 1 - 2 135 71
BI856275.1 - 1 - 2 182 71

View Full Product Details