The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015659.2 | NP_056474.2 | 6 - 7 | 797 | 85 |
GenBank mRNA | AF083127.1 | - | 5 - 6 | 500 | 85 |
AJ007398.1 | - | 6 - 7 | 729 | 85 | |
AK000822.1 | - | 6 - 7 | 741 | 85 | |
AK295935.1 | - | 5 - 6 | 652 | 85 | |
AL049999.1 | - | 6 - 7 | 783 | 85 | |
AY154473.1 | - | 6 - 7 | 758 | 85 | |
AY598331.1 | - | 6 - 7 | 724 | 85 | |
BC017899.1 | - | 6 - 7 | 746 | 85 | |
BC019069.2 | - | 6 - 7 | 730 | 85 | |
BC070325.1 | - | 6 - 7 | 748 | 85 | |
BC107782.1 | - | 6 - 7 | 750 | 85 | |
BC112228.1 | - | 6 - 7 | 762 | 85 | |
BC113699.1 | - | 6 - 7 | 750 | 85 |