Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A28
Entrez Gene ID
81894
Gene Name
solute carrier family 25 member 28
Gene Aliases
MFRN2, MRS3/4, MRS4L, NPD016
Chromosome Location
Chr.10: 99610518 - 99659329 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031212.3 NP_112489.3 3 - 4 712 81
XM_005270208.1 XP_005270265.1 3 - 4 702 81
XM_005270211.4 XP_005270268.1 3 - 4 316 81
XM_006718005.3 XP_006718068.1 3 - 4 376 81
XM_011540239.2 XP_011538541.1 3 - 4 598 81
XM_011540240.1 XP_011538542.1 4 - 5 1848 81
XM_017016741.1 XP_016872230.1 3 - 4 1784 81
XM_017016742.1 XP_016872231.1 3 - 4 1605 81
GenBank mRNA AF327402.1 - 3 - 4 612 81
AF327403.1 - 3 - 4 1053 81
AJ303077.1 - 3 - 4 612 81
AJ303078.1 - 3 - 4 1053 81
BC047312.1 - 3 - 4 659 81
BC058937.1 - 3 - 4 621 81
BC076399.1 - 3 - 4 652 81
BC094821.1 - 3 - 4 395 81

View Full Product Details