The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014146.3 | NP_054865.2 | 4 - 5 | 543 | 92 |
NM_032463.2 | NP_115852.1 | 5 - 6 | 662 | 92 | |
NM_032464.2 | NP_115853.2 | 5 - 6 | 732 | 92 | |
XM_011516558.2 | XP_011514860.1 | 5 - 6 | 572 | 92 | |
GenBank mRNA | AF161531.1 | - | 4 - 5 | 295 | 92 |
AF252611.1 | - | 5 - 6 | 483 | 92 | |
AF252612.1 | - | 4 - 5 | 294 | 92 | |
AF252613.1 | - | 4 - 5 | 279 | 92 | |
AF252614.1 | - | 5 - 6 | 413 | 92 | |
AF257135.1 | - | 3 - 4 | 214 | 92 | |
AK092904.1 | - | 5 - 6 | 873 | 92 | |
AK225150.1 | - | 4 - 5 | 294 | 92 | |
AK290074.1 | - | 5 - 6 | 410 | 92 | |
AK290916.1 | - | 4 - 5 | 291 | 92 | |
AY190023.1 | - | 3 - 4 | 224 | 92 | |
BC001609.1 | - | 5 - 6 | 544 | 92 | |
BC009204.2 | - | 3 - 4 | 216 | 92 |