Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLRN1
Entrez Gene ID
7401
Gene Name
clarin 1
Gene Aliases
RP61, USH3, USH3A
Chromosome Location
Chr.3: 150926163 - 150972999 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052995.2 NP_443721.1 3 - 4 734 100
GenBank mRNA AF388366.1 - 3 - 4 734 100

View Full Product Details