Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LAT2
- Entrez Gene ID
-
7462
- Gene Name
- linker for activation of T-cells family member 2
- Gene Aliases
-
HSPC046, LAB, NTAL, WBSCR15, WBSCR5, WSCR5
- Chromosome Location
-
Chr.7: 74209757 - 74229834 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF252612.1 |
- |
7 - 8 |
417 |
72 |
View Full Product Details