The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004523.3 | NP_004514.2 | 12 - 13 | 1804 | 119 |
GenBank mRNA | AK314258.1 | - | 12 - 13 | 1707 | 119 |
BC126211.1 | - | 12 - 13 | 1603 | 119 | |
BC136474.1 | - | 12 - 13 | 1603 | 119 | |
U37426.1 | - | 12 - 13 | 1586 | 119 | |
X85137.1 | - | 12 - 13 | 1506 | 119 |