Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KIF11
Entrez Gene ID
3832
Gene Name
kinesin family member 11
Gene Aliases
EG5, HKSP, KNSL1, MCLMR, TRIP5
Chromosome Location
Chr.10: 92593068 - 92655395 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004523.3 NP_004514.2 13 - 14 2009 104
GenBank mRNA AK314258.1 - 13 - 14 1912 104
BC126211.1 - 13 - 14 1808 104
BC136474.1 - 13 - 14 1808 104
U37426.1 - 13 - 14 1791 104
X85137.1 - 13 - 14 1711 104

View Full Product Details