The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004523.3 | NP_004514.2 | 18 - 19 | 2861 | 96 |
GenBank mRNA | AB208964.1 | - | 4 - 5 | 596 | 96 |
AK314258.1 | - | 18 - 19 | 2764 | 96 | |
BC126211.1 | - | 18 - 19 | 2660 | 96 | |
BC136474.1 | - | 18 - 19 | 2660 | 96 | |
U37426.1 | - | 18 - 19 | 2643 | 96 | |
X85137.1 | - | 18 - 19 | 2566 | 96 |