The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002311.4 | NP_002302.2 | 12 - 13 | 2037 | 69 |
NM_013975.3 | NP_039269.2 | 12 - 13 | 2037 | 69 | |
XM_005257970.3 | XP_005258027.1 | 12 - 13 | 2005 | 69 | |
XM_006721896.3 | XP_006721959.1 | 12 - 13 | 2006 | 69 | |
XM_017024624.1 | XP_016880113.1 | 12 - 13 | 2201 | 69 | |
GenBank mRNA | AK300363.1 | - | 12 - 13 | 1997 | 69 |
BC068005.1 | - | 12 - 13 | 2012 | 69 | |
U40671.1 | - | 12 - 13 | 1998 | 69 | |
X84740.1 | - | 12 - 13 | 1980 | 69 |