The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002311.4 | NP_002302.2 | 7 - 8 | 1417 | 124 |
NM_013975.3 | NP_039269.2 | 7 - 8 | 1417 | 124 | |
XM_005257970.3 | XP_005258027.1 | 7 - 8 | 1385 | 124 | |
XM_006721896.3 | XP_006721959.1 | 7 - 8 | 1386 | 124 | |
XM_017024624.1 | XP_016880113.1 | 7 - 8 | 1581 | 124 | |
GenBank mRNA | AK300363.1 | - | 7 - 8 | 1377 | 124 |
AK308407.1 | - | 7 - 8 | 1362 | 124 | |
BC068005.1 | - | 7 - 8 | 1392 | 124 | |
U40671.1 | - | 7 - 8 | 1378 | 124 | |
X84740.1 | - | 7 - 8 | 1360 | 124 |