Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ECM2
Entrez Gene ID
1842
Gene Name
extracellular matrix protein 2
Gene Aliases
-
Chromosome Location
Chr.9: 92493547 - 92536092 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001197295.1 NP_001184224.1 1 - 2 162 166
NM_001197296.1 NP_001184225.1 1 - 2 162 166
NM_001393.3 NP_001384.1 1 - 2 162 166
GenBank mRNA AB011792.1 - 1 - 2 48 166
AK296458.1 - 1 - 2 65 166
AK298278.1 - 2 - 3 74 166
AK303922.1 - 1 - 2 63 166
AK312820.1 - 1 - 2 60 166
BX537976.1 - 1 - 2 125 166

View Full Product Details