The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242803.1 | NP_001229732.1 | 4 - 5 | 582 | 67 |
NM_006994.4 | NP_008925.1 | 9 - 10 | 1237 | 67 | |
NM_197974.2 | NP_932078.2 | 8 - 9 | 1145 | 67 | |
GenBank mRNA | AB209581.1 | - | 5 - 6 | 2765 | 67 |
AK223279.1 | - | 9 - 10 | 1172 | 67 | |
AK223301.1 | - | 9 - 10 | 1156 | 67 | |
AK291722.1 | - | 9 - 10 | 1221 | 67 | |
AK301553.1 | - | 8 - 9 | 1129 | 67 | |
BC015815.2 | - | 9 - 10 | 1174 | 67 | |
BT007251.1 | - | 7 - 8 | 994 | 67 | |
BX341863.2 | - | 4 - 5 | 498 | 67 | |
KM244652.1 | - | 4 - 5 | 490 | 67 | |
U90548.1 | - | 9 - 10 | 1165 | 67 |