The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290312.1 | NP_001277241.1 | 1 - 2 | 514 | 120 |
NM_001324001.1 | NP_001310930.1 | 1 - 2 | 514 | 120 | |
NM_001324002.1 | NP_001310931.1 | 1 - 2 | 514 | 120 | |
NM_001324009.1 | NP_001310938.1 | 1 - 2 | 514 | 120 | |
NM_001324013.1 | NP_001310942.1 | 1 - 2 | 514 | 120 | |
GenBank mRNA | BC031262.1 | - | 1 - 2 | 93 | 120 |
DA349847.1 | - | 1 - 2 | 514 | 120 | |
HQ589335.1 | - | 1 - 2 | 348 | 120 |