The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018037.1 | NP_001018047.1 | 53 - 54 | 7792 | 117 |
NM_001018038.2 | NP_001018048.1 | 54 - 55 | 7909 | 117 | |
NM_015186.3 | NP_056001.1 | 54 - 55 | 7909 | 117 | |
NM_033305.2 | NP_150648.2 | 54 - 55 | 7909 | 117 | |
GenBank mRNA | AB023203.1 | - | 15 - 16 | 2740 | 117 |
AB054005.1 | - | 54 - 55 | 7820 | 117 | |
AF337532.1 | - | 54 - 55 | 7909 | 117 | |
AJ608769.1 | - | 53 - 54 | 7792 | 117 | |
AJ626859.1 | - | 54 - 55 | 7909 | 117 | |
AK022967.1 | - | 7 - 8 | 1067 | 117 | |
AK027023.1 | - | 1 - 2 | 211 | 117 | |
BC041852.1 | - | 12 - 13 | 2104 | 117 |